siRNA Details
Gene Symbol: Arap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178407.3 | NP_848494.2 | 2 | 911 | ||
GenBank mRNA | AK037027.1 | - | 2 | 820 | ||
AK039153.1 | - | 2 | 829 | |||
AK041540.1 | - | 2 | 854 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
