siRNA Details
Gene Symbol: 9930021J03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172836.3 | NP_766424.2 | 8 | 6150 | ||
GenBank mRNA | AK036879.1 | - | 1 | 1260 | ||
AK220440.1 | - | 1 | 1481 | |||
BC125582.1 | - | 1 | 1281 | 40130097 |
