siRNA Details
Gene Symbol: Ccnj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172839.4 | NP_766427.1 | 6 | 1282 | ||
GenBank mRNA | AK052506.1 | - | 6 | 1283 | ||
AK157841.1 | - | 6 | 1276 | |||
BC050857.1 | - | 3 | 733 | |||
BC117559.1 | - | 5 | 1753 | 6517343 | ||
BC120921.2 | - | 6 | 1264 | 40093200 | ||
BC120922.1 | - | 6 | 1264 | 40093202 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
