siRNA Details
Gene Symbol: St18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244692.1 | NP_001231621.1 | 8 | 1346 | ||
NM_001244693.1 | NP_001231622.1 | 7 | 1268 | |||
NM_173868.2 | NP_776293.1 | 6 | 1159 | |||
GenBank mRNA | AB097467.1 | - | 6 | 1184 | ||
AJ536155.1 | - | 8 | 1310 | |||
AK032557.1 | - | 8 | 1311 | |||
AK122303.1 | - | 6 | 1108 | |||
AK133743.1 | - | 8 | 1276 | |||
AK137440.1 | - | 6 | 1109 | |||
AK141513.1 | - | 6 | 1068 | |||
AK147320.1 | - | 8 | 1347 | |||
AK147664.1 | - | 8 | 1347 | |||
BC032273.1 | - | 1 | 73 | |||
BC118528.1 | - | 7 | 1182 | 40098452 |
