siRNA Details
Gene Symbol: Plekha6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160268.1 | NP_001153740.1 | 8, 9 | 1217 | ||
NM_182930.2 | NP_891846.1 | 8, 9 | 1157 | |||
GenBank mRNA | AK149273.1 | - | 1, 2 | 485 | ||
AK173074.1 | - | 8, 9 | 1188 | |||
BC038061.1 | - | 7, 8 | 1154 | |||
BC054547.1 | - | 8, 9 | 1148 | 5709381 |
