siRNA Details
Gene Symbol: Rgsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243223.1 | NP_001230152.1 | 15 | 2651 | ||
GenBank mRNA | AK029630.1 | - | 6 | 758 | ||
AK161463.1 | - | 8 | 1071 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
