siRNA Details
Gene Symbol: Mettl11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143956.1 | NP_001137428.1 | 2 | 459 | ||
GenBank mRNA | BC157906.1 | - | 2 | 459 | 9088108 | |
BC158115.1 | - | 2 | 436 | 9088188 | ||
BC172048.1 | - | 2 | 459 | 9054334 | ||
BC172056.1 | - | 3 | 517 |
