siRNA Details
Gene Symbol: Mettl11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143956.1 | NP_001137428.1 | 3 | 651 | ||
GenBank mRNA | BC157906.1 | - | 3 | 651 | 9088108 | |
BC158115.1 | - | 3 | 628 | 9088188 | ||
BC172048.1 | - | 3 | 651 | 9054334 | ||
BC172056.1 | - | 4 | 620 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
