siRNA Details
Gene Symbol: Scyl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286002.1 | NP_001272931.1 | 7 | 1003 | ||
NM_001286003.1 | NP_001272932.1 | 7 | 1215 | |||
NM_028776.5 | NP_083052.1 | 7 | 1190 | |||
GenBank mRNA | AK004809.2 | - | 7 | 930 | ||
AK030824.1 | - | 7 | 1165 | |||
AK044100.1 | - | 7 | 1176 | |||
AK162937.1 | - | 7 | 1004 | |||
AK184387.1 | - | 1 | 83 | |||
BC043085.1 | - | 7 | 891 | 6417440 | ||
BC066800.1 | - | 7 | 875 |

More Information
Other Species Targeted

Gene Ontology Categories:
