siRNA Details
Gene Symbol: Kif26b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161665.1 | NP_001155137.1 | 2 | 708 | ||
GenBank mRNA | AB355846.1 | - | 2 | 708 | ||
AK052046.1 | - | 2 | 709 | |||
BC120791.1 | - | 2 | 371 | 40129714 | ||
BC137689.1 | - | 2 | 371 | 8860708 |
