siRNA Details
Gene Symbol: Noxa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163626.1 | NP_001157098.1 | 5 | 845 | ||
NM_172204.4 | NP_757341.3 | 5 | 845 | |||
GenBank mRNA | AF539798.1 | - | 5 | 564 | ||
AK087263.1 | - | 5 | 771 | |||
AY255770.1 | - | 5 | 564 | |||
BC047532.1 | - | 5 | 836 | 4988389 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
