siRNA Details
Gene Symbol: Ppp1r26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005420.1 | NP_001005420.1 | 3 | 1108 | ||
GenBank mRNA | AK147361.1 | - | 1 | 3704 | ||
AK172993.1 | - | 1 | 866 | |||
BC070440.1 | - | 3 | 1108 | 30362439 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
