siRNA Details
Gene Symbol: Fam78a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175511.4 | NP_780720.1 | 2 | 1406 | ||
GenBank mRNA | AK077626.1 | - | 2 | 1411 | ||
AK079514.1 | - | 2 | 1407 | |||
AK165375.1 | - | 1774 | ||||
BC137951.1 | - | 2 | 900 | 8860970 | ||
BC137952.1 | - | 2 | 900 | 8860971 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
