siRNA Details
Gene Symbol: Ralgps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290570.1 | NP_001277499.1 | 8 | 911 | ||
NM_001290572.1 | NP_001277501.1 | 9 | 962 | |||
NM_175211.5 | NP_780420.1 | 9 | 962 | |||
GenBank mRNA | AK076767.1 | - | 9 | 947 | ||
AK129123.1 | - | 8 | 878 | |||
AK170248.1 | - | 9 | 909 | |||
BC072656.1 | - | 9 | 897 | 30536315 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
