siRNA Details
Gene Symbol: Zbtb34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085507.1 | NP_001078976.1 | 3 | 1647 | ||
NM_001276332.1 | NP_001263261.1 | 2 | 1527 | |||
GenBank mRNA | AK173321.1 | - | 2 | 1520 |
