siRNA Details
Gene Symbol: Pde11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081033.1 | NP_001074502.1 | 20 | 2705 | ||
GenBank mRNA | AK050924.1 | - | 2 | 76 | ||
AK192697.1 | - | 2 | 77 | |||
BC158125.1 | - | 20 | 2845 | 9088197 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
