siRNA Details
Gene Symbol: Slc5a12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003915.2 | NP_001003915.1 | 12 | 1726 | ||
NM_001177624.1 | NP_001171095.1 | 12 | 1714 | |||
GenBank mRNA | AF529222.1 | - | 12 | 1734 | ||
AK085356.1 | - | 12 | 1709 | |||
AK128951.1 | - | 12 | 1706 | |||
AK144004.1 | - | 12 | 1647 | |||
AK165419.1 | - | 12 | 1707 | |||
AY964639.1 | - | 12 | 1417 | |||
BC121791.1 | - | 12 | 1410 | |||
BC141255.1 | - | 12 | 1642 | 9055867 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
