siRNA Details
Gene Symbol: Rpl27a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011975.3 | NP_036105.2 | 3 | 157 | ||
GenBank mRNA | AK007837.1 | - | 3 | 156 | ||
AK008531.1 | - | 3 | 155 | |||
AK010286.1 | - | 3 | 158 | |||
AK011361.1 | - | 3 | 157 | |||
AK138110.1 | - | 2 | 426 | |||
AK150907.1 | - | 3 | 155 | |||
AK167792.1 | - | 3 | 156 | |||
AK167915.1 | - | 3 | 156 | |||
AK213005.1 | - | 43 | ||||
BC056958.1 | - | 2 | 141 | 6817520 | ||
BC081430.1 | - | 2 | 141 | 6411497 | ||
BC086939.1 | - | 3 | 154 | 6760797 | ||
CJ072009.1 | - | 3 | 159 | |||
X05021.1 | - | 3 | 154 |

More Information
Other Species Targeted

Gene Ontology Categories:
