siRNA Details
Gene Symbol: Ppm1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178726.3 | NP_848841.2 | 1 | 782 | ||
GenBank mRNA | AK032529.1 | - | 1 | 840 | ||
AK035912.1 | - | 1 | 785 | |||
AK045724.1 | - | 1 | 823 | |||
AK131646.1 | - | 1 | 769 | |||
AK142789.1 | - | 1 | 800 | |||
AK147389.1 | - | 1 | 785 | |||
AK147876.1 | - | 1 | 819 | |||
AK165160.1 | - | 1 | 815 | |||
AK220523.1 | - | 1 | 787 | |||
AY184801.1 | - | 1 | 370 | |||
BC096031.1 | - | 1 | 782 | 6837066 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
