siRNA Details
Gene Symbol: Ppm1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178726.3 | NP_848841.2 | 3 | 1235 | ||
GenBank mRNA | AK028275.1 | - | 3 | 277 | ||
AK032529.1 | - | 3 | 1293 | |||
AK035912.1 | - | 3 | 1238 | |||
AK045724.1 | - | 3 | 1276 | |||
AK131646.1 | - | 3 | 1222 | |||
AK142789.1 | - | 3 | 1253 | |||
AK147389.1 | - | 3 | 1236 | |||
AK147876.1 | - | 3 | 1272 | |||
AK165160.1 | - | 3 | 1268 | |||
AK220523.1 | - | 3 | 1240 | |||
AY184801.1 | - | 3 | 823 | |||
BC096031.1 | - | 3 | 1235 | 6837066 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
