siRNA Details
Gene Symbol: Zfp697 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172863.4 | NP_766451.3 | 2 | 193 | ||
GenBank mRNA | BC092534.1 | - | 2 | 126 | 30249621 | |
BQ890991.1 | - | 2 | 112 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
