siRNA Details
Gene Symbol: Slc22a15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039371.2 | NP_001034460.2 | 6 | 1259 | ||
GenBank mRNA | AK029695.1 | - | 3 | 417 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc22a15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039371.2 | NP_001034460.2 | 6 | 1259 | ||
GenBank mRNA | AK029695.1 | - | 3 | 417 |