siRNA Details
Gene Symbol: Slc44a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081263.1 | NP_001074732.1 | 20 | 2244 | ||
GenBank mRNA | BC068154.1 | - | 7 | 1029 | ||
BC086641.1 | - | 20 | 2244 | 6417957 |
