siRNA Details
Gene Symbol: Lrrc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081358.2 | NP_001074827.1 | 25 | 5598 | ||
NM_001291452.1 | NP_001278381.1 | 24 | 5457 | |||
NM_001291453.1 | NP_001278382.1 | 23 | 5337 | |||
GenBank mRNA | AK122498.1 | - | 24 | 4592 | ||
BC138450.1 | - | 23 | 5244 | 8861470 |

More Information
Other Species Targeted

Gene Ontology Categories:
