siRNA Details
Gene Symbol: Atp6v0d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175406.3 | NP_780615.2 | 2, 3 | 382 | ||
GenBank mRNA | AB088358.1 | - | 2, 3 | 338 | ||
AK075745.1 | - | 2, 3 | 422 | |||
AK089507.1 | - | 2, 3 | 307 | |||
AK137586.1 | - | 2, 3 | 362 | |||
AK170072.1 | - | 2, 3 | 356 | |||
AK170405.1 | - | 2, 3 | 355 | |||
AK170578.1 | - | 2, 3 | 326 | |||
AK170659.1 | - | 2, 3 | 331 | |||
AY145896.1 | - | 2, 3 | 382 | |||
AY517482.1 | - | 2, 3 | 374 | |||
BC087899.1 | - | 2, 3 | 316 | 30504725 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
