siRNA Details
Gene Symbol: Manea | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172865.2 | NP_766453.2 | 5 | 1355 | ||
GenBank mRNA | AK030141.1 | - | 5 | 1330 | ||
BC067076.1 | - | 5 | 1355 | 30628555 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
