siRNA Details
Gene Symbol: Slc1a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146255.2 | NP_666367.2 | 2 | 410 | ||
GenBank mRNA | AK044586.1 | - | 2 | 302 | ||
AK044671.1 | - | 2 | 340 | |||
BC030400.1 | - | 2 | 421 | 5363389 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
