siRNA Details
Gene Symbol: Ifnlr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174851.3 | NP_777276.3 | 5 | 691 | ||
GenBank mRNA | AK141338.1 | - | 5 | 692 | ||
AY184376.1 | - | 5 | 606 | |||
BC057856.1 | - | 5 | 624 | 5036481 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
