siRNA Details
Gene Symbol: Lrrc38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162983.1 | NP_001156455.1 | 1 | 60 | ||
GenBank mRNA | AK038658.1 | - | 1 | 22 | ||
AK078190.1 | - | 1 | 34 | |||
BC129963.1 | - | 1 | 53 | |||
BC129964.1 | - | 1 | 60 |

More Information
Other Species Targeted

Gene Ontology Categories:
