siRNA Details
Gene Symbol: Ptchd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083342.1 | NP_001076811.1 | 2 | 1126 | ||
GenBank mRNA | AB235901.1 | - | 1 | 880 | ||
AK090326.1 | - | 2 | 1110 | |||
AK173152.1 | - | 1 | 569 | |||
BC070441.1 | - | 2 | 1058 | 30535318 | ||
BC072569.1 | - | 2 | 1091 | 5719275 |
