siRNA Details
Gene Symbol: Ptchd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083342.1 | NP_001076811.1 | 13 | 2774 | ||
GenBank mRNA | AB235901.1 | - | 12 | 2528 | ||
AK078196.1 | - | 3 | 278 | |||
AK090326.1 | - | 13 | 2795 | |||
AK173152.1 | - | 12 | 2217 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_003750087.
Gene Ontology Categories:
