siRNA Details
Gene Symbol: Ptchd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083342.1 | NP_001076811.1 | 14 | 3029 | ||
GenBank mRNA | AB235901.1 | - | 13 | 2783 | ||
AK078196.1 | - | 4 | 533 | |||
AK090326.1 | - | 14 | 3050 | |||
AK173152.1 | - | 13 | 2472 | |||
BC072569.1 | - | 14 | 2933 | 5719275 |
