siRNA Details
Gene Symbol: Actr3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004365.1 | NP_001004365.1 | 8 | 774 | ||
GenBank mRNA | AK035798.1 | - | 7 | 686 | ||
BC053106.1 | - | 7 | 747 | |||
BC082279.1 | - | 8 | 774 | 6332588 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
