siRNA Details
Gene Symbol: Kctd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175519.5 | NP_780728.3 | 2 | 1659 | ||
GenBank mRNA | AK039167.1 | - | 2 | 1629 | ||
AK042569.1 | - | 2 | 1515 | |||
AK043351.1 | - | 2 | 1629 | |||
AK045439.1 | - | 2 | 1512 | |||
AK047519.1 | - | 2 | 1660 | |||
AY615967.1 | - | 2 | 1661 | |||
BC116940.1 | - | 2 | 1310 | 40126259 | ||
BC116942.1 | - | 2 | 1310 | 40126261 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
