siRNA Details
Gene Symbol: Tmprss11f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178730.3 | NP_848845.1 | 3 | 213 | ||
GenBank mRNA | AK028447.1 | - | 3 | 215 | ||
AK028587.1 | - | 3 | 208 | |||
AK029032.1 | - | 3 | 175 | |||
AK037173.1 | - | 2 | 57 | |||
BC132629.1 | - | 2 | 63 | 40130906 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
