siRNA Details
Gene Symbol: Hcar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175520.4 | NP_780729.2 | 1 | 1223 | ||
GenBank mRNA | AK029064.1 | - | 1 | 1208 | ||
AK046700.1 | - | 1 | 1225 | |||
BC141305.1 | - | 1 | 1094 | 9055917 | ||
EU809459.1 | - | 1 | 829 | |||
EU809460.1 | - | 1 | 805 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
