siRNA Details
Gene Symbol: Lrrc61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110160.1 | NP_001103630.1 | 2 | 1831 | ||
NM_177736.3 | NP_808404.2 | 3 | 2060 | |||
GenBank mRNA | AK143222.1 | - | 3 | 2063 | ||
AK171463.1 | - | 2 | 1831 | |||
BC027309.1 | - | 4 | 1861 | 3981816 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
