siRNA Details
Gene Symbol: Zfp775 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173429.2 | NP_775605.1 | 2 | 922 | ||
GenBank mRNA | AK048054.1 | - | 2 | 922 | ||
AK163249.1 | - | 3 | 1028 | |||
AK167452.1 | - | 3 | 1028 | |||
BC141223.1 | - | 2 | 917 | 9055835 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
