siRNA Details
Gene Symbol: Doxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029987.1 | NP_001025158.1 | 1 | 1139 | ||
GenBank mRNA | BC150935.1 | - | 1 | 1208 | 9087846 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Doxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029987.1 | NP_001025158.1 | 1 | 1139 | ||
GenBank mRNA | BC150935.1 | - | 1 | 1208 | 9087846 |