siRNA Details
Gene Symbol: Igkv1-135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BF301241.1 | - | 260 | |||
M17722.1 | - | 2 | 217 | |||
M20830.1 | - | 2 | 255 | |||
M91696.1 | - | 1 | 183 | |||
U30241.1 | - | 1 | 134 |
Gene Symbol: Igkc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC092090.1 | - | 229 | |||
Z22077.1 | - | 163 |
Gene Symbol: Igkv1-133 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | Z17401.1 | - | 2 | 235 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
