siRNA Details
Gene Symbol: 2010107G12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025573.2 | NP_001020744.2 | 6 | 916 | ||
GenBank mRNA | BC075651.1 | - | 6 | 916 | 30541436 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 2010107G12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025573.2 | NP_001020744.2 | 6 | 916 | ||
GenBank mRNA | BC075651.1 | - | 6 | 916 | 30541436 |