siRNA Details
Gene Symbol: E330009J07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175528.4 | NP_780737.1 | 4 | 661 | ||
GenBank mRNA | AK054277.1 | - | 4 | 662 | ||
AK122457.1 | - | 3 | 359 | |||
AK145280.1 | - | 4 | 653 | |||
AK147402.1 | - | 4 | 654 |
