siRNA Details
Gene Symbol: Fkrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173430.2 | NP_775606.1 | 3 | 291 | ||
GenBank mRNA | AJ511806.1 | - | 3 | 274 | ||
AK080624.1 | - | 1 | 100 | |||
AK135089.1 | - | 3 | 292 | |||
AK156156.1 | - | 3 | 289 | |||
AK166068.1 | - | 3 | 293 | |||
AK168308.1 | - | 3 | 289 | |||
BC029216.1 | - | 3 | 286 | |||
BC053072.1 | - | 3 | 274 | 5686286 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
