siRNA Details
Gene Symbol: Ggn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182694.2 | NP_874353.2 | 2 | 2392 | ||
NM_182696.2 | NP_874355.1 | 2 | 348 | |||
GenBank mRNA | AF538032.1 | - | 2 | 1881 | ||
AF538033.1 | - | 3 | 1301 | |||
AF538034.1 | - | 2 | 348 | |||
BC089358.1 | - | 2 | 2392 | 30610755 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
