siRNA Details
Gene Symbol: Lgi4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144556.2 | NP_653139.2 | 5 | 506 | ||
GenBank mRNA | AJ487521.1 | - | 5 | 507 | ||
AJ487960.1 | - | 5 | 506 | |||
AK033967.1 | - | 5 | 324 | |||
AK134643.1 | - | 5 | 1100 | |||
AK148793.1 | - | 5 | 765 |

More Information
Other Species Targeted

Gene Ontology Categories:
