siRNA Details
Gene Symbol: Lgi4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144556.2 | NP_653139.2 | 3 | 347 | ||
GenBank mRNA | AJ487521.1 | - | 3 | 348 | ||
AJ487960.1 | - | 3 | 347 | |||
AK033967.1 | - | 3 | 165 | |||
AK134643.1 | - | 3 | 941 | |||
AK148793.1 | - | 3 | 606 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
