siRNA Details
Gene Symbol: Tshz3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172298.2 | NP_758502.1 | 2 | 3170 | ||
GenBank mRNA | AK037176.1 | - | 1 | 493 | ||
AK041793.1 | - | 1 | 2056 | |||
AK086984.1 | - | 1 | 1553 | |||
AK220396.1 | - | 2 | 3041 | |||
AY063491.1 | - | 2 | 3170 | |||
BC150674.1 | - | 2 | 3113 | 9087585 |

More Information
Other Species Targeted

Gene Ontology Categories:
