siRNA Details
Gene Symbol: 4930433I11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207248.3 | NP_997131.2 | 2 | 1379 | ||
GenBank mRNA | AK029662.1 | - | 2 | 1435 | ||
BC150916.1 | - | 2 | 1379 | 9087827 | ||
BC150921.1 | - | 2 | 1379 | 9087832 |
