siRNA Details
Gene Symbol: Siglecg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK036698.1 | - | 4 | 896 | ||
AK156766.1 | - | 4 | 924 | |||
AK162797.1 | - | 3 | 827 | |||
BC114981.1 | - | 4 | 887 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
