siRNA Details
Gene Symbol: Mrgprx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034868.3 | NP_001030040.2 | 2 | 912 | ||
GenBank mRNA | AK148121.1 | - | 2 | 843 | ||
AK148319.1 | - | 1 | 233 | |||
BC141225.1 | - | 2 | 835 | 9055837 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
